| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023041.hg.1 |
ENST00000336926.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023041.hg.1 |
ENST00000434438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023041.hg.1 |
NM_001243198.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023041.hg.1 |
NM_005338.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023041.hg.1 |
TC07001534.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
ENST00000336926.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
ENST00000434438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
ENST00000616821.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
NM_001243198.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
NM_005338.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
ENST00000336926.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
ENST00000434438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
ENST00000616821.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
NM_001243198.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
NM_005338.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700182218.hg.1 |
TC0700011556.hg.1 |
Clariom D TC |