Assay Details
| Gene Symbol: RTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206900.2 | NP_996783.1 | 4 - 5 | 1047 | 81 | |
| GenBank mRNA | AF004223.1 | - | 2 - 3 | 315 | 81 | |
| AF038540.1 | - | 2 - 3 | 496 | 81 | ||
| AK291967.1 | - | 4 - 5 | 1028 | 81 | ||
| AK314158.1 | - | 4 - 5 | 1030 | 81 | ||
More Information
Set Membership: |
|

RTN2
Genomic Map