Assay Details
| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015245.2 | NP_056060.2 | 3 - 4 | 572 | 61 | |
| XM_011514434.2 | XP_011512736.1 | 3 - 4 | 560 | 61 | ||
| XM_011514435.2 | XP_011512737.1 | 3 - 4 | 560 | 61 | ||
| GenBank mRNA | AK298987.1 | - | 3 - 4 | 530 | 61 | |
| BC022396.1 | - | 3 - 4 | 572 | 61 | ||
| BC031934.1 | - | 4 - 5 | 644 | 4819948 | 61 | |
| BC132832.1 | - | 3 - 4 | 509 | 40146854 | 61 | |
| D86982.1 | - | 3 - 4 | 569 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000510.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC031934.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06005355.hg.1 | ENST00000360359.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005355.hg.1 | ENST00000373990.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005355.hg.1 | ENST00000535627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005355.hg.1 | ENST00000544150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005355.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005355.hg.1 | TC06000510.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153877.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153877.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153877.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153877.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153877.hg.1 | TC0600007774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600153875.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153875.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153875.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153875.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153875.hg.1 | TC0600007774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000360359.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | ENST00000470698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007774.hg.1 | NM_015245.2 | Refseq |

ANKS1A
Genomic Map