Assay Details
| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC031934.1 | - | 24 - 25 | 3108 | 4819948 | 86 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKS1A| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC031934.1 | - | 24 - 25 | 3108 | 4819948 | 86 |
Set Membership: |
|