Assay Details
| Gene Symbol: SLC28A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004212.3 | NP_004203.2 | 15 - 16 | 1713 | 137 | |
| XM_011522198.2 | XP_011520500.1 | 15 - 16 | 1733 | 137 | ||
| XM_011522202.2 | XP_011520504.1 | 7 - 8 | 884 | 137 | ||
| GenBank mRNA | AF036109.1 | - | 14 - 15 | 1662 | 137 | |
| AK222709.1 | - | 15 - 16 | 1713 | 137 | ||
| AK291974.1 | - | 15 - 16 | 1684 | 137 | ||
| BC093737.1 | - | 14 - 15 | 1676 | 7939582 | 137 | |
| BC112047.1 | - | 14 - 15 | 1688 | 8327515 | 137 | |
| U84392.1 | - | 15 - 16 | 1707 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000354.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15003231.hg.1 | ENST00000347644.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003231.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003231.hg.1 | TC15000354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003233.hg.1 | ENST00000347644.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003233.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003233.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003233.hg.1 | TC15000354.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150633.hg.1 | TC1500007116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500150631.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150631.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150631.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150631.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150631.hg.1 | TC1500007116.hg.1 | Clariom D TC |

SLC28A2
Genomic Map