Assay Details
| Gene Symbol: SLC28A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004212.3 | NP_004203.2 | 1 - 2 | 44 | 64 | |
| GenBank mRNA | AK222709.1 | - | 1 - 2 | 44 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000354.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_004212.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15003218.hg.1 | ENST00000347644.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003218.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003218.hg.1 | TC15000354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003216.hg.1 | ENST00000347644.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003216.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003216.hg.1 | TC15000354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150615.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150615.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150615.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150615.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150615.hg.1 | TC1500007116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500150612.hg.1 | TC1500007116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000347644.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000559924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | ENST00000560767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007116.hg.1 | NM_004212.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 206104 | - | - |

SLC28A2
Genomic Map