Assay Details
| Gene Symbol: GPR108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017027012.1 | XP_016882501.1 | 15 - 16 | 1355 | 85 | |
| XM_017027014.1 | XP_016882503.1 | 15 - 16 | 1355 | 85 | ||
| XM_017027015.1 | XP_016882504.1 | 15 - 16 | 1543 | 85 | ||
More Information
Set Membership: |
|

GPR108
Genomic Map