Assay Details
| Gene Symbol: CHAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005483.2 | NP_005474.2 | 1 - 2 | 158 | 151 | |
| XM_011527605.2 | XP_011525907.1 | 1 - 2 | 225 | 151 | ||
| XM_011527606.2 | XP_011525908.1 | 1 - 2 | 225 | 151 | ||
| XM_011527607.2 | XP_011525909.1 | 1 - 2 | 225 | 151 | ||
| GenBank mRNA | BC052620.1 | - | 1 - 2 | 157 | 151 | |
| BC067093.1 | - | 1 - 2 | 159 | 6339027 | 151 | |
| BQ889365.1 | - | 1 - 2 | 158 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000088.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC052620.1,BC067093.1,NM_005483.2,XM_011527605.2,XM_011527606.2,XM_011527607.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19001407.hg.1 | ENST00000301280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001407.hg.1 | ENST00000344143.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001407.hg.1 | ENST00000535117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001407.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001407.hg.1 | TC19000088.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001408.hg.1 | ENST00000301280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001408.hg.1 | ENST00000344143.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001408.hg.1 | ENST00000535117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001408.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001408.hg.1 | TC19000088.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900146964.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146964.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146964.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146964.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146964.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146966.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146968.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000587739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | ENST00000589648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006691.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | NM_005483.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | TC1900006691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | ENST00000301280.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | ENST00000585854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | ENST00000587580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146967.hg.1 | NM_005483.2 | Refseq |

CHAF1A
Genomic Map