Assay Details
| Gene Symbol: HSPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130106.1 | NP_001123578.1 | 3 - 4 | 547 | 68 | |
| NM_001297600.1 | NP_001284529.1 | 1 - 2 | 462 | 68 | ||
| NM_012267.4 | NP_036399.3 | 2 - 3 | 656 | 68 | ||
| XM_005258700.2 | XP_005258757.1 | 2 - 3 | 387 | 68 | ||
| XM_005258701.2 | XP_005258758.1 | 2 - 3 | 449 | 68 | ||
| XM_011526691.1 | XP_011524993.1 | 3 - 4 | 545 | 68 | ||
| XM_017026543.1 | XP_016882032.1 | 2 - 3 | 384 | 68 | ||
| XM_017026544.1 | XP_016882033.1 | 2 - 3 | 384 | 68 | ||
| XM_017026545.1 | XP_016882034.1 | 2 - 3 | 387 | 68 | ||
| GenBank mRNA | AB020592.1 | - | 2 - 3 | 378 | 68 | |
| AF093420.1 | - | 2 - 3 | 381 | 68 | ||
| AF187859.1 | - | 2 - 3 | 346 | 68 | ||
| AF217996.1 | - | 3 - 4 | 533 | 68 | ||
| AK075293.1 | - | 2 - 3 | 403 | 68 | ||
| AK129755.1 | - | 2 - 3 | 459 | 68 | ||
| AK130636.1 | - | 409 | 68 | |||
| AK130781.1 | - | 2 - 3 | 393 | 68 | ||
| AK294358.1 | - | 1 - 2 | 461 | 68 | ||
| BC001236.1 | - | 1 - 2 | 311 | 3454675 | 68 | |
| BC002373.2 | - | 2 - 3 | 368 | 2822836 | 68 | |
| CR457118.1 | - | 1 - 2 | 213 | 68 | ||
More Information
Set Membership: |
|

HSPBP1
Genomic Map