Assay Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184726.1 | NP_001171655.1 | 3 - 4 | 599 | 87 | |
| GenBank mRNA | BC011239.1 | - | 3 - 4 | 478 | 4153362 | 87 |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000471659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000471659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000471659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000215838.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000405742.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000407817.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000423350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000450638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000471659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000471659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000471659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | ENST00000493542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_000355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009257.hg.1 | NM_001184726.1 | Refseq |

TCN2
Genomic Map