Assay Details
| Gene Symbol: NYNRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025081.2 | NP_079357.2 | 5 - 6 | 2861 | 64 | |
| GenBank mRNA | AB037726.2 | - | 3 - 4 | 2306 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000169.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14001911.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001911.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001911.hg.1 | TC14000169.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001907.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001907.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001907.hg.1 | TC14000169.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001909.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001909.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001909.hg.1 | TC14000169.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001910.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001910.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001910.hg.1 | TC14000169.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001912.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001912.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001912.hg.1 | TC14000169.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001908.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001908.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001908.hg.1 | TC14000169.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400146871.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146871.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146871.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146871.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146871.hg.1 | TC1400006741.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400146868.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146868.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146868.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146868.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146868.hg.1 | TC1400006741.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400146870.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146870.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146870.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146870.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146870.hg.1 | TC1400006741.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400146874.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146874.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146874.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146874.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146874.hg.1 | TC1400006741.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400146873.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146873.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146873.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146873.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146873.hg.1 | TC1400006741.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400146872.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146872.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146872.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146872.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146872.hg.1 | TC1400006741.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000382554.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | ENST00000554505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006741.hg.1 | NM_025081.2 | Refseq |

NYNRIN
Genomic Map