Assay Details
| Gene Symbol: WDFY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014991.4 | NP_055806.2 | 38 - 39 | 6642 | 94 | |
| XM_005262858.4 | XP_005262915.1 | 40 - 41 | 6683 | 94 | ||
| XM_011531757.2 | XP_011530059.1 | 39 - 40 | 6594 | 94 | ||
| XM_011531759.2 | XP_011530061.1 | 38 - 39 | 6556 | 94 | ||
| XM_011531760.2 | XP_011530062.1 | 38 - 39 | 6494 | 94 | ||
| XM_011531761.2 | XP_011530063.1 | 39 - 40 | 6549 | 94 | ||
| XM_011531762.2 | XP_011530064.1 | 39 - 40 | 6594 | 94 | ||
| XM_011531763.2 | XP_011530065.1 | 37 - 38 | 6440 | 94 | ||
| XM_011531764.2 | XP_011530066.1 | 38 - 39 | 6510 | 94 | ||
| XM_011531765.2 | XP_011530067.1 | 36 - 37 | 6296 | 94 | ||
| XM_011531766.2 | XP_011530068.1 | 39 - 40 | 6594 | 94 | ||
| XM_017007906.1 | XP_016863395.1 | 41 - 42 | 6811 | 94 | ||
| XM_017007907.1 | XP_016863396.1 | 41 - 42 | 6808 | 94 | ||
| XM_017007908.1 | XP_016863397.1 | 40 - 41 | 6740 | 94 | ||
| XM_017007909.1 | XP_016863398.1 | 26 - 27 | 4436 | 94 | ||
| GenBank mRNA | AB023210.2 | - | 3 - 4 | 325 | 94 | |
| AF538685.1 | - | 37 - 38 | 6534 | 94 | ||
| BC150246.1 | - | 3 - 4 | 346 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001355.hg.1
Set Membership: |
|

WDFY3
Genomic Map