Assay Details
| Gene Symbol: TTC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318533.1 | NP_001305462.1 | 8 - 9 | 690 | 101 | |
| NM_017868.3 | NP_060338.3 | 8 - 9 | 672 | 101 | ||
| XM_005271604.3 | XP_005271661.1 | 8 - 9 | 888 | 101 | ||
| XM_011542890.2 | XP_011541192.1 | 9 - 10 | 886 | 101 | ||
| XM_017017943.1 | XP_016873432.1 | 8 - 9 | 672 | 101 | ||
| XM_017017944.1 | XP_016873433.1 | 9 - 10 | 1242 | 101 | ||
| XM_017017945.1 | XP_016873434.1 | 9 - 10 | 725 | 101 | ||
| XM_017017946.1 | XP_016873435.1 | 8 - 9 | 895 | 101 | ||
| XM_017017947.1 | XP_016873436.1 | 8 - 9 | 672 | 101 | ||
| XM_017017948.1 | XP_016873437.1 | 8 - 9 | 672 | 101 | ||
| XM_017017949.1 | XP_016873438.1 | 8 - 9 | 672 | 101 | ||
| XM_017017950.1 | XP_016873439.1 | 8 - 9 | 672 | 101 | ||
| GenBank mRNA | AK000542.1 | - | 7 - 8 | 564 | 101 | |
| AK023921.1 | - | 8 - 9 | 618 | 101 | ||
| AK091887.1 | - | 10 - 11 | 1284 | 101 | ||
| AK223117.1 | - | 7 - 8 | 564 | 101 | ||
| AK292331.1 | - | 8 - 9 | 610 | 101 | ||
| BC011546.1 | - | 7 - 8 | 614 | 101 | ||
| BC032355.1 | - | 7 - 8 | 594 | 5219117 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

TTC12
Genomic Map