Assay Details
| Gene Symbol: ZMYND8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005260360.4 | XP_005260417.1 | 1 - 2 | 736 | 104 | |
| XM_005260364.2 | XP_005260421.1 | 1 - 2 | 71 | 104 | ||
| XM_017027764.1 | XP_016883253.1 | 1 - 2 | 681 | 104 | ||
| XM_017027767.1 | XP_016883256.1 | 1 - 2 | 826 | 104 | ||
| XM_017027769.1 | XP_016883258.1 | 1 - 2 | 71 | 104 | ||
| XM_017027771.1 | XP_016883260.1 | 1 - 2 | 71 | 104 | ||
| XM_017027776.1 | XP_016883265.1 | 1 - 2 | 71 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005260360.4, XM_005260364.2, XM_017027764.1, XM_017027767.1, XM_017027769.1, XM_017027771.1, XM_017027776.1 transcripts.
Set Membership: |
|

ZMYND8
Genomic Map