| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000385275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009550.hg.1 |
NR_029668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012176.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012176.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012176.hg.1 |
TC08000955.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012178.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012178.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012178.hg.1 |
TC08000955.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165110.hg.1 |
TC0800009550.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800165108.hg.1 |
TC0800009550.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000385275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000517675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000518557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000519461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
ENST00000521863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_024281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009550.hg.1 |
NR_029668.1 |
Refseq |