Assay Details
| Gene Symbol: SORCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014978.2 | NP_055793.1 | 24 - 25 | 3570 | 147 | |
| XM_011539542.2 | XP_011537844.1 | 19 - 20 | 2321 | 147 | ||
| GenBank mRNA | AB028982.1 | - | 24 - 25 | 3570 | 147 | |
| BC045754.1 | - | 4 - 5 | 475 | 147 | ||
| BC117417.1 | - | 24 - 25 | 3386 | 40125968 | 147 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000788.hg.1
Set Membership: |
|

SORCS3
Genomic Map