Assay Details
| Gene Symbol: SORCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014978.2 | NP_055793.1 | 8 - 9 | 1527 | 70 | |
| XM_011539542.2 | XP_011537844.1 | 3 - 4 | 278 | 70 | ||
| GenBank mRNA | AB028982.1 | - | 8 - 9 | 1527 | 70 | |
| BC117417.1 | - | 8 - 9 | 1343 | 40125968 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000788.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009398.hg.1 | ENST00000369701.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009398.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009398.hg.1 | TC10000788.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | ENST00000393176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008813.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | ENST00000369699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | ENST00000369701.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | NM_014978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160325.hg.1 | TC1000008813.hg.1 | Clariom D TC |

SORCS3
Genomic Map