Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100128988 | LOC100128988 | -12.5kb | Sense | Chr. 20 - 40980761 - 41025375 on NCBI Build GRCh38 | |
| Downstream | PLCG1 antisense RNA 1 | PLCG1-AS1 | TOP1-AS1 | 60.2kb | Anti-sense | Chr. 20 - 41097994 - 41138000 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map