Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031438.3 | NP_113626.1 | 2 - 3 | 313 | 75 | |
| XM_005272095.1 | XP_005272152.1 | 2 - 3 | 298 | 75 | ||
| GenBank mRNA | AK098066.1 | - | 2 - 3 | 282 | 75 | |
| AL136592.1 | - | 2 - 3 | 268 | 75 | ||
| BC041099.1 | - | 2 - 3 | 297 | 5495144 | 75 | |
| CB131118.1 | - | 2 - 3 | 301 | 75 | ||
| DB029218.1 | - | 2 - 3 | 315 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001649.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000515407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000515407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | ENST00000515407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022396.hg.1 | ENST00000230792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022396.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022396.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022396.hg.1 | TC05001649.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022398.hg.1 | ENST00000230792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022398.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022398.hg.1 | TC05001649.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022395.hg.1 | ENST00000230792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022395.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022395.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022395.hg.1 | TC05001649.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000515407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000515407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | ENST00000515407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011614.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180425.hg.1 | TC0500011614.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | ENST00000507423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | NM_001300741.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180426.hg.1 | TC0500011614.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | ENST00000230792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | ENST00000508889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | NM_031438.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180428.hg.1 | TC0500011614.hg.1 | Clariom D TC |

NUDT12
Genomic Map