Assay Details
| Gene Symbol: HSPA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005346.4 | NP_005337.2 | 1 | 169 | 127 | |
| GenBank mRNA | AK295398.1 | - | 1 | 167 | 127 | |
| AK303967.1 | - | 1 | 167 | 127 | ||
| BC001876.1 | - | 1 | 157 | 127 | ||
| BC057397.1 | - | 1 | 145 | 6188453 | 127 | |
| BC063507.1 | - | 1 | 151 | 4153196 | 127 | |
| DB015158.1 | - | 1 | 169 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000385.hg.1, TC6_apd_hap1000040.hg.1, TC6_cox_hap2000079.hg.1, TC6_dbb_hap3000070.hg.1, TC6_qbl_hap6000070.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK295398.1,AK303967.1,BC057397.1,BC063507.1,NM_005346.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of gene expression
regulation of cell death
regulation of protein ubiquitination
negative regulation of protein ubiquitination
positive regulation of interleukin-8 production
cellular response to oxidative stress
cellular response to heat
protein refolding
ATP metabolic process
viral entry into host cell
protein stabilization
positive regulation of NF-kappaB transcription factor activity
negative regulation of cell death
cellular heat acclimation
positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
negative regulation of inclusion body assembly
regulation of cellular response to heat
positive regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
virus receptor activity
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
histone deacetylase binding
protein binding involved in protein folding
unfolded protein binding
C3HC4-type RING finger domain binding
positive regulation of gene expression
regulation of cell death
regulation of protein ubiquitination
negative regulation of protein ubiquitination
positive regulation of interleukin-8 production
cellular response to oxidative stress
cellular response to heat
protein refolding
ATP metabolic process
viral entry into host cell
protein stabilization
positive regulation of NF-kappaB transcription factor activity
negative regulation of cell death
cellular heat acclimation
positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
negative regulation of inclusion body assembly
regulation of cellular response to heat
positive regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
virus receptor activity
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
histone deacetylase binding
protein binding involved in protein folding
unfolded protein binding
C3HC4-type RING finger domain binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001279.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001279.hg.1 | TC6_cox_hap2000079.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001220.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001220.hg.1 | TC6_dbb_hap3000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001233.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001233.hg.1 | TC6_qbl_hap6000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001234.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001234.hg.1 | TC6_qbl_hap6000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003961.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003961.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003961.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003961.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | ENST00000418382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | ENST00000545241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | ENST00000545429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003966.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000508.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000508.hg.1 | TC6_apd_hap1000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001221.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001221.hg.1 | TC6_dbb_hap3000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003967.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003967.hg.1 | ENST00000418382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003967.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003967.hg.1 | ENST00000545429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003967.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003967.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000506.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000506.hg.1 | TC6_apd_hap1000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001235.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001235.hg.1 | TC6_qbl_hap6000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003962.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003962.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003962.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003962.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000507.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000507.hg.1 | TC6_apd_hap1000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000509.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000509.hg.1 | TC6_apd_hap1000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001278.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001278.hg.1 | TC6_cox_hap2000079.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003968.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003968.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003968.hg.1 | ENST00000545429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003968.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003968.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003969.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003969.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003969.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003969.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001222.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001222.hg.1 | TC6_dbb_hap3000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001223.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001223.hg.1 | TC6_dbb_hap3000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001232.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001232.hg.1 | TC6_qbl_hap6000070.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003963.hg.1 | ENST00000375650.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003963.hg.1 | ENST00000542758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003963.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06003963.hg.1 | TC06000385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001277.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001277.hg.1 | TC6_cox_hap2000079.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001280.hg.1 | NM_005346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001280.hg.1 | TC6_cox_hap2000079.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152124.hg.1 | TC0600007613.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600152126.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152126.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152126.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152126.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152126.hg.1 | TC0600007613.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007613.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | ENST00000375651.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | ENST00000608703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | NM_005345.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600152125.hg.1 | TC0600007613.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600152137.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152137.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152137.hg.1 | TC0600007616.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600152139.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152139.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152139.hg.1 | TC0600007616.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007616.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152136.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152136.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152136.hg.1 | TC0600007616.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600152138.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152138.hg.1 | ENST00000375650.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600152138.hg.1 | TC0600007616.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148242 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 233284 | - | - |

HSPA1B
Genomic Map