Assay Details
| Gene Symbol: WDR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032259.3 | NP_115635.1 | 7 - 8 | 2802 | 63 | |
| XM_011522699.2 | XP_011521001.1 | 8 - 9 | 3306 | 63 | ||
| GenBank mRNA | AL136863.1 | - | 7 - 8 | 2779 | 63 | |
| BC008025.1 | - | 7 - 8 | 2267 | 3544791 | 63 | |
| BC009761.2 | - | 7 - 8 | 1802 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000735.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16010656.hg.1 | ENST00000248142.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010656.hg.1 | ENST00000293883.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010656.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16010656.hg.1 | TC16000735.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | TC1600008972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600167721.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000248142.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000293883.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | ENST00000567014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008972.hg.1 | NM_032259.3 | Refseq |

WDR24
Genomic Map