| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
ENST00000251722.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
ENST00000427112.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
ENST00000435959.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
ENST00000443711.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
ENST00000450966.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
NM_018218.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046424.hg.1 |
TC02002882.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
ENST00000251722.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
ENST00000427112.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
ENST00000435959.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
ENST00000443711.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
ENST00000450966.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
NM_018218.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046423.hg.1 |
TC02002882.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000251722.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000427112.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000427947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000443711.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000450966.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000484528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
NM_018218.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000251722.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000427112.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000427947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000443711.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000450966.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
ENST00000484528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
NM_018218.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218214.hg.1 |
TC0200016093.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000251722.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000427112.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000427947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000443711.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000450966.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000484528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
NM_018218.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000251722.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000427112.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000427947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000443711.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000450966.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
ENST00000484528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
NM_018218.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218213.hg.1 |
TC0200016093.hg.1 |
Clariom D TC |