Assay Details
| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032223.3 | NP_115599.2 | 13 - 14 | 2550 | 65 | |
| XM_005273987.4 | XP_005274044.1 | 13 - 14 | 4470 | 65 | ||
| XM_005273990.4 | XP_005274047.1 | 13 - 14 | 4473 | 65 | ||
| XM_006718549.3 | XP_006718612.1 | 13 - 14 | 4455 | 65 | ||
| XM_006718551.3 | XP_006718614.1 | 13 - 14 | 4473 | 65 | ||
| XM_011545024.2 | XP_011543326.1 | 14 - 15 | 3646 | 65 | ||
| XM_011545025.2 | XP_011543327.1 | 13 - 14 | 4473 | 65 | ||
| XM_011545026.2 | XP_011543328.1 | 7 - 8 | 1313 | 65 | ||
| XM_011545027.1 | XP_011543329.1 | 5 - 6 | 526 | 65 | ||
| XM_011545028.2 | XP_011543330.1 | 13 - 14 | 4473 | 65 | ||
| GenBank mRNA | CN262095.1 | - | 2 - 3 | 160 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000635.hg.1
Set Membership: |
|

PCNX3
Genomic Map