Assay Details
| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032223.3 | NP_115599.2 | 4 - 5 | 483 | 92 | |
| XM_005273987.4 | XP_005274044.1 | 4 - 5 | 2367 | 92 | ||
| XM_005273990.4 | XP_005274047.1 | 4 - 5 | 2367 | 92 | ||
| XM_006718549.3 | XP_006718612.1 | 4 - 5 | 2367 | 92 | ||
| XM_006718551.3 | XP_006718614.1 | 4 - 5 | 2367 | 92 | ||
| XM_011545024.2 | XP_011543326.1 | 5 - 6 | 1540 | 92 | ||
| XM_011545025.2 | XP_011543327.1 | 4 - 5 | 2367 | 92 | ||
| XM_011545028.2 | XP_011543330.1 | 4 - 5 | 2367 | 92 | ||
| GenBank mRNA | DY654455.1 | - | 4 - 5 | 380 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000635.hg.1
Set Membership: |
|

PCNX3
Genomic Map