Assay Details
| Gene Symbol: ATG4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032852.3 | NP_116241.2 | 8 - 9 | 1249 | 84 | |
| NM_178221.2 | NP_835739.1 | 8 - 9 | 1201 | 84 | ||
| XM_005271288.2 | XP_005271345.1 | 8 - 9 | 1192 | 84 | ||
| XM_011542313.2 | XP_011540615.1 | 8 - 9 | 1223 | 84 | ||
| XM_017002600.1 | XP_016858089.1 | 8 - 9 | 1223 | 84 | ||
| XM_017002601.1 | XP_016858090.1 | 8 - 9 | 1201 | 84 | ||
| GenBank mRNA | AJ312234.1 | - | 7 - 8 | 1054 | 84 | |
| AJ320169.1 | - | 7 - 8 | 1058 | 84 | ||
| AK027773.1 | - | 8 - 9 | 1220 | 84 | ||
| BC008395.1 | - | 8 - 9 | 1146 | 84 | ||
| BC018678.1 | - | 8 - 9 | 1207 | 84 | ||
| BC033024.1 | - | 8 - 9 | 1208 | 4838543 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000710.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011361.hg.1 | ENST00000317868.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011361.hg.1 | ENST00000371120.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011361.hg.1 | ENST00000540025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011361.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011361.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011361.hg.1 | TC01000710.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | ENST00000443289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008564.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | ENST00000317868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | ENST00000371120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | ENST00000414558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | NM_032852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | NM_178221.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160795.hg.1 | TC0100008564.hg.1 | Clariom D TC |

ATG4C
Genomic Map