Assay Details
| Gene Symbol: ANKRD52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173595.3 | NP_775866.2 | 4 - 5 | 355 | 90 | |
| XM_011538197.2 | XP_011536499.1 | 3 - 4 | 457 | 90 | ||
| XM_017019183.1 | XP_016874672.1 | 3 - 4 | 538 | 90 | ||
| XM_017019184.1 | XP_016874673.1 | 4 - 5 | 387 | 90 | ||
| GenBank mRNA | AB190990.1 | - | 4 - 5 | 372 | 90 | |
| AK125379.1 | - | 4 - 5 | 372 | 90 | ||
| DR002390.1 | - | 4 - 5 | 363 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ANKRD52
Genomic Map