Assay Details
| Gene Symbol: PADI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007365.2 | NP_031391.2 | 11 - 12 | 1388 | 132 | |
| XM_017000148.1 | XP_016855637.1 | 3 - 4 | 562 | 132 | ||
| GenBank mRNA | AB023211.1 | - | 11 - 12 | 1370 | 132 | |
| AB030176.1 | - | 11 - 12 | 1370 | 132 | ||
| AK127006.1 | - | 3 - 4 | 933 | 132 | ||
| AK295783.1 | - | 8 - 9 | 1040 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002287.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin organization
regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding
chromatin organization
regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01034863.hg.1 | ENST00000375486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034863.hg.1 | ENST00000444885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034863.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034863.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034863.hg.1 | TC01002287.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | ENST00000479534.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013076.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | ENST00000375486.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | ENST00000466151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | NM_007365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196651.hg.1 | TC0100013076.hg.1 | Clariom D TC |

PADI2
Genomic Map