Assay Details
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146686.2 | NP_001140158.1 | 1 - 2 | 61 | 59 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002111.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001146686.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037319.hg.1 | ENST00000442080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037319.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037319.hg.1 | TC03002111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037321.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037321.hg.1 | TC03002111.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000479491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | ENST00000495042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013528.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300200917.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300200917.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300200917.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300200917.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300200917.hg.1 | TC0300013528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | ENST00000442080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | ENST00000456552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | NM_001146686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300200915.hg.1 | TC0300013528.hg.1 | Clariom D TC |

GMNC
Genomic Map