Assay Details
| Gene Symbol: FBXL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105043.1 | 3 - 4 | 341 | 134 | ||
| XM_005250205.3 | XP_005250262.1 | 3 - 4 | 333 | 134 | ||
| XM_005250207.3 | XP_005250264.1 | 3 - 4 | 333 | 134 | ||
| XM_005250208.3 | XP_005250265.1 | 3 - 4 | 333 | 134 | ||
| XM_011515928.2 | XP_011514230.1 | 3 - 4 | 333 | 134 | ||
| XM_011515929.2 | XP_011514231.1 | 3 - 4 | 333 | 134 | ||
| XM_011515932.2 | XP_011514234.1 | 3 - 4 | 333 | 134 | ||
| XM_017011850.1 | XP_016867339.1 | 3 - 4 | 277 | 134 | ||
| XM_017011851.1 | XP_016867340.1 | 3 - 4 | 333 | 134 | ||
| XM_017011852.1 | XP_016867341.1 | 3 - 4 | 483 | 134 | ||
| GenBank mRNA | BC026121.1 | - | 3 - 4 | 344 | 4826546 | 134 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

FBXL13
Genomic Map