Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 6 like | TAF6L | PAF65A | Anti-sense | Chr. 11 - 62771303 - 62787342 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 179B | TMEM179B | 80.0 | Anti-sense | Chr. 11 - 62787321 - 62790400 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map