Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270639.1 | NP_001257568.1 | 3 - 4 | 525 | 123 | |
| NM_001270640.1 | NP_001257569.1 | 3 - 4 | 522 | 123 | ||
| NM_001270686.1 | NP_001257615.1 | 3 - 4 | 368 | 123 | ||
| NM_138334.3 | NP_612207.1 | 3 - 4 | 371 | 123 | ||
| XM_011526434.2 | XP_011524736.1 | 3 - 4 | 365 | 123 | ||
| GenBank mRNA | AF247787.1 | - | 2 - 3 | 300 | 123 | |
| BC051380.1 | - | 3 - 4 | 329 | 123 | ||
| BC062416.1 | - | 3 - 4 | 348 | 6503623 | 123 | |
| BM970016.1 | - | 2 - 3 | 123 | |||
| BP328576.1 | - | 3 - 4 | 368 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000595718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000595718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011207.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_138334.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | TC1900011207.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000594350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000595669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000598418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000601423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | ENST00000602146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270639.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270640.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270641.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_001270686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194100.hg.1 | NM_138334.3 | Refseq |

JOSD2
Genomic Map