Assay Details
| Gene Symbol: DCHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003737.3 | NP_003728.1 | 12 - 13 | 5570 | 80 | |
| GenBank mRNA | AB053446.1 | - | 12 - 13 | 5573 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001363.hg.1
Set Membership: |
|

DCHS1
Genomic Map