| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11017168.hg.1 |
ENST00000299441.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11017168.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11017168.hg.1 |
TC11001363.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175456.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100175456.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175456.hg.1 |
ENST00000299441.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100175456.hg.1 |
NM_003737.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100175456.hg.1 |
TC1100009982.hg.1 |
Clariom D TC |