Assay Details
| Gene Symbol: SLC6A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020208.3 | NP_064593.1 | 5 - 6 | 823 | 76 | |
| XM_011533847.2 | XP_011532149.1 | 4 - 5 | 1137 | 76 | ||
| XM_011533848.2 | XP_011532150.1 | 5 - 6 | 833 | 76 | ||
| GenBank mRNA | AF075260.1 | - | 5 - 6 | 701 | 76 | |
| AF125107.1 | - | 6 - 7 | 887 | 76 | ||
| AJ276207.1 | - | 5 - 6 | 750 | 76 | ||
| BC126197.1 | - | 5 - 6 | 763 | 8991913 | 76 | |
| BC136431.1 | - | 5 - 6 | 763 | 9020420 | 76 | |
| BC144087.1 | - | 5 - 6 | 763 | 9052607 | 76 | |
More Information
Set Membership: |
|

SLC6A20
Genomic Map