| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008184.hg.1 |
ENST00000254950.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008184.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16008184.hg.1 |
TC16000567.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
TC1600011421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191167.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
TC1600011421.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600191169.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000254950.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000562754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000564399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
ENST00000566354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011421.hg.1 |
NM_013245.2 |
Refseq |