| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
ENST00000335166.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
ENST00000353139.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
ENST00000360679.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
ENST00000394406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
ENST00000427330.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
ENST00000452074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
NM_198834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
NM_198836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
NM_198837.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
NM_198838.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
NM_198839.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17018619.hg.1 |
TC17001406.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000612895.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000613146.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000614428.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000616317.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000617649.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198837.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198838.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198839.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
TC1700010488.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000612895.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000613146.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000614428.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000616317.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
ENST00000617649.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198837.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198838.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700182427.hg.1 |
NM_198839.2 |
Refseq |