Assay Details
| Gene Symbol: ACACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198834.2 | NP_942131.1 | 29 - 30 | 4200 | 70 | |
| NM_198836.2 | NP_942133.1 | 29 - 30 | 3768 | 70 | ||
| NM_198837.1 | NP_942134.1 | 27 - 28 | 3838 | 70 | ||
| NM_198838.1 | NP_942135.1 | 28 - 29 | 3949 | 70 | ||
| NM_198839.2 | NP_942136.1 | 33 - 34 | 4874 | 70 | ||
| XM_005257267.4 | XP_005257324.1 | 29 - 30 | 3681 | 70 | ||
| XM_011524701.1 | XP_011523003.1 | 29 - 30 | 4121 | 70 | ||
| XM_011524703.1 | XP_011523005.1 | 29 - 30 | 3753 | 70 | ||
| XM_011524704.2 | XP_011523006.1 | 29 - 30 | 3686 | 70 | ||
| XM_017024553.1 | XP_016880042.1 | 30 - 31 | 4235 | 70 | ||
| XM_017024555.1 | XP_016880044.1 | 29 - 30 | 3834 | 70 | ||
| GenBank mRNA | AB371587.1 | - | 29 - 30 | 3747 | 70 | |
| AY237919.1 | - | 27 - 28 | 3577 | 70 | ||
| AY315627.1 | - | 27 - 28 | 3577 | 70 | ||
| BC137287.1 | - | 29 - 30 | 3705 | 9021284 | 70 | |
| U19822.1 | - | 27 - 28 | 3577 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ACACA
Genomic Map