Assay Details
| Gene Symbol: SH3PXD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014631.2 | NP_055446.2 | 6 - 7 | 571 | 69 | |
| XM_005270294.4 | XP_005270351.1 | 6 - 7 | 620 | 69 | ||
| XM_006718079.3 | XP_006718142.1 | 6 - 7 | 624 | 69 | ||
| XM_017016969.1 | XP_016872458.1 | 6 - 7 | 629 | 69 | ||
| XM_017016970.1 | XP_016872459.1 | 4 - 5 | 361 | 69 | ||
| GenBank mRNA | AK056469.1 | - | 5 - 6 | 294 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001635.hg.1
Set Membership: |
|

SH3PXD2A
Genomic Map