Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005247569.2 | XP_005247626.1 | 2 - 3 | 276 | 84 | |
| XM_017006715.1 | XP_016862204.1 | 2 - 3 | 436 | 84 | ||
| GenBank mRNA | BC040355.1 | - | 2 - 3 | 249 | 4748104 | 84 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC040355.1,XM_005247569.2,XM_017006715.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158071.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158071.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158071.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158071.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158071.hg.1 | TC0300008148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300158072.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158072.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158072.hg.1 | TC0300008148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158073.hg.1 | TC0300008148.hg.1 | Clariom D TC |

TMEM45A
Genomic Map