Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018004.2 | NP_060474.1 | 2 - 3 | 499 | 117 | |
| XM_005247569.2 | XP_005247626.1 | 3 - 4 | 460 | 117 | ||
| XM_017006715.1 | XP_016862204.1 | 3 - 4 | 620 | 117 | ||
| GenBank mRNA | AB013911.1 | - | 1 - 2 | 186 | 117 | |
| AK000996.1 | - | 2 - 3 | 499 | 117 | ||
| AK129789.1 | - | 2 - 3 | 349 | 117 | ||
| AY450392.1 | - | 2 - 3 | 499 | 117 | ||
| BC040355.1 | - | 3 - 4 | 433 | 4748104 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000509.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009578.hg.1 | ENST00000323523.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009578.hg.1 | ENST00000403410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009578.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009578.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009578.hg.1 | TC03000509.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000485260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000488904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | ENST00000489060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008148.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000323523.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000403410.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000449609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | ENST00000462884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | NM_018004.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158076.hg.1 | TC0300008148.hg.1 | Clariom D TC |

TMEM45A
Genomic Map