Assay Details
| Gene Symbol: RIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272077.1 | NP_001259006.1 | 1 - 2 | 279 | 132 | |
| NM_002930.3 | NP_002921.1 | 1 - 2 | 279 | 132 | ||
| GenBank mRNA | AF493922.1 | - | 1 - 2 | 132 | ||
| AK313823.1 | - | 1 - 2 | 184 | 132 | ||
| AY563951.1 | - | 1 - 2 | 106 | 132 | ||
| BC018060.1 | - | 1 - 2 | 250 | 4816017 | 132 | |
| U71204.1 | - | 1 - 2 | 279 | 132 | ||
| U78164.1 | - | 1 - 2 | 278 | 132 | ||
| U78166.1 | - | 1 - 2 | 278 | 132 | ||
| Y07565.1 | - | 1 - 2 | 280 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000470.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
small GTPase mediated signal transduction
Ras protein signal transduction
chemical synaptic transmission
positive regulation of neuron projection development
regulation of endocytosis
regulation of Cdc42 protein signal transduction
maintenance of protein location in cell
intracellular signal transduction
positive regulation of MAPK cascade
regulation of calcium-mediated signaling
GTPase activity
protein binding
calmodulin binding
GTP binding
semaphorin receptor binding
identical protein binding
MAPK cascade
small GTPase mediated signal transduction
Ras protein signal transduction
chemical synaptic transmission
positive regulation of neuron projection development
regulation of endocytosis
regulation of Cdc42 protein signal transduction
maintenance of protein location in cell
intracellular signal transduction
positive regulation of MAPK cascade
regulation of calcium-mediated signaling
GTPase activity
protein binding
calmodulin binding
GTP binding
semaphorin receptor binding
identical protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18004817.hg.1 | ENST00000282028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004817.hg.1 | ENST00000326695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004817.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004817.hg.1 | TC18000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004816.hg.1 | ENST00000282028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004816.hg.1 | ENST00000326695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004816.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004816.hg.1 | TC18000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004818.hg.1 | ENST00000282028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004818.hg.1 | ENST00000326695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004818.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004818.hg.1 | TC18000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000594852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | ENST00000594852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | TC1800008549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158241.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | TC1800008549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158243.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000594852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | ENST00000594852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008549.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | TC1800008549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158245.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | NM_002930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | TC1800008549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | ENST00000326695.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | ENST00000589109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | ENST00000590910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | NM_001272077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158242.hg.1 | NM_002930.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124859 | - | - |

RIT2
Genomic Map