Assay Details
| Gene Symbol: MRPL39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080794.3 | NP_542984.2 | 8 - 9 | 934 | 144 | |
| XM_006724026.3 | XP_006724089.1 | 8 - 9 | 926 | 144 | ||
| GenBank mRNA | AF270511.1 | - | 5 - 6 | 473 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial genome maintenance
tRNA aminoacylation for protein translation
prolyl-tRNA aminoacylation
biological_process
mitochondrial translational elongation
mitochondrial translational termination
nucleotide binding
RNA binding
proline-tRNA ligase activity
ligase activity, forming aminoacyl-tRNA and related compounds
poly(A) RNA binding
mitochondrial genome maintenance
tRNA aminoacylation for protein translation
prolyl-tRNA aminoacylation
biological_process
mitochondrial translational elongation
mitochondrial translational termination
nucleotide binding
RNA binding
proline-tRNA ligase activity
ligase activity, forming aminoacyl-tRNA and related compounds
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003794.hg.1 | ENST00000307301.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003794.hg.1 | ENST00000352957.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003794.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003794.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003794.hg.1 | TC21000338.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000307301.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000352957.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | ENST00000419219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_017446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007796.hg.1 | NM_080794.3 | Refseq |

MRPL39
Genomic Map