Assay Details
| Gene Symbol: RUNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015051.3 | NP_001015051.3 | 7 - 8 | 1234 | 98 | |
| NM_001278478.1 | NP_001265407.1 | 5 - 6 | 982 | 98 | ||
| XM_011514962.2 | XP_011513264.1 | 7 - 8 | 1409 | 98 | ||
| GenBank mRNA | BC108919.1 | - | 5 - 6 | 964 | 98 | |
| BC108920.1 | - | 5 - 6 | 968 | 98 | ||
| L40992.1 | - | 5 - 6 | 847 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000621.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ossification
osteoblast differentiation
endochondral ossification
osteoblast fate commitment
chondrocyte differentiation
chondrocyte development
osteoblast development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
hemopoiesis
neuron differentiation
T cell differentiation
BMP signaling pathway
positive regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
regulation of fibroblast growth factor receptor signaling pathway
odontogenesis of dentin-containing tooth
regulation of odontogenesis of dentin-containing tooth
cellular response to fibroblast growth factor stimulus
regulation of cell differentiation
positive regulation of osteoblast differentiation
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
cell maturation
embryonic cranial skeleton morphogenesis
stem cell differentiation
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
ATP binding
protein domain specific binding
histone deacetylase binding
bHLH transcription factor binding
repressing transcription factor binding
ossification
osteoblast differentiation
endochondral ossification
osteoblast fate commitment
chondrocyte differentiation
chondrocyte development
osteoblast development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
hemopoiesis
neuron differentiation
T cell differentiation
BMP signaling pathway
positive regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
regulation of fibroblast growth factor receptor signaling pathway
odontogenesis of dentin-containing tooth
regulation of odontogenesis of dentin-containing tooth
cellular response to fibroblast growth factor stimulus
regulation of cell differentiation
positive regulation of osteoblast differentiation
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
cell maturation
embryonic cranial skeleton morphogenesis
stem cell differentiation
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
ATP binding
protein domain specific binding
histone deacetylase binding
bHLH transcription factor binding
repressing transcription factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000359524.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371436.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000371438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000465038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000473041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000473041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000478660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000483377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000576263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | ENST00000625924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001015051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001024630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NM_001278478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103532.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103532.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103532.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103532.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103532.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103532.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008146.hg.1 | NR_103533.1 | Refseq |

RUNX2
Genomic Map