Assay Details
| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015397.3 | NP_056212.1 | 1 - 2 | 416 | 153 | |
| GenBank mRNA | AB067479.1 | - | 1 - 2 | 336 | 153 | |
| AK292935.1 | - | 1 - 2 | 222 | 153 | ||
| BC063823.1 | - | 1 - 2 | 334 | 6184796 | 153 | |
| DQ518909.1 | - | 1 - 2 | 336 | 153 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001028.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000466402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000466402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | ENST00000466402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013496.hg.1 | ENST00000361264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013496.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013496.hg.1 | TC09001028.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013492.hg.1 | ENST00000361264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013492.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013492.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013492.hg.1 | TC09001028.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013497.hg.1 | ENST00000361264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013497.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013497.hg.1 | TC09001028.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000466402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000466402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | ENST00000466402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009882.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167747.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167747.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167747.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167747.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167747.hg.1 | TC0900009882.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167746.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167746.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167746.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167746.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167746.hg.1 | TC0900009882.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000396990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000450964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | ENST00000463286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167741.hg.1 | TC0900009882.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167745.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167745.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167745.hg.1 | ENST00000361264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167745.hg.1 | NM_015397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167745.hg.1 | TC0900009882.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 209246 | - | - |

DCAF12
Genomic Map