| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000343523.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000357033.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000359836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000361471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378677.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378680.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378682.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378684.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378702.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378707.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000378723.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000474231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000534884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000535280.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000541735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
ENST00000542849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_000109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004006.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004009.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004010.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004013.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004014.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004015.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004016.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004017.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004018.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004020.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
NM_004023.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012572.hg.1 |
TC0X000943.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000343523.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000357033.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000358062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000359836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000361471.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378677.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378680.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378702.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378707.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378723.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000474231.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000541735.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000619831.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000620040.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_000109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004006.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004009.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004010.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004013.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004014.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004015.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004016.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004017.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004020.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004023.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000343523.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000357033.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000358062.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000359836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000361471.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378677.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378680.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378702.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378707.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000378723.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000474231.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000541735.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000619831.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
ENST00000620040.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_000109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004006.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004009.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004010.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004013.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004014.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004015.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004016.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004017.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004020.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
NM_004023.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163957.hg.1 |
TC0X00009341.hg.1 |
Clariom D TC |