Assay Details
| Gene Symbol: PRODH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021232.1 | NP_067055.1 | 7 - 8 | 1083 | 119 | |
| XM_017027084.1 | XP_016882573.1 | 6 - 7 | 940 | 119 | ||
| XM_017027085.1 | XP_016882574.1 | 6 - 7 | 939 | 119 | ||
| GenBank mRNA | BC039710.1 | - | 6 - 7 | 855 | 119 | |
| U80018.1 | - | 7 - 8 | 1083 | 119 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001455.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019660.hg.1 | ENST00000301175.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019660.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019660.hg.1 | TC19001455.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | TC1900010499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185951.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | NM_021232.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | TC1900010499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | ENST00000301175.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | ENST00000587695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | ENST00000588266.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185949.hg.1 | NM_021232.1 | Refseq |

PRODH2
Genomic Map