Assay Details
| Gene Symbol: LINC01114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090794.1 | - | 1 | 657 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232270 | - | - |

LINC01114
Genomic Map