| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000483055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000483055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000483055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
ENST00000360670.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
ENST00000372179.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
ENST00000393228.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
ENST00000467453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009713.hg.1 |
TC09000755.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000483055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000483055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000483055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
ENST00000490694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009018.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
ENST00000372179.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
ENST00000393229.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
NM_032536.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900161381.hg.1 |
TC0900009018.hg.1 |
Clariom D TC |