Assay Details
| Gene Symbol: GDPGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013657.2 | NP_001013679.2 | 4 | 1305 | 67 | |
| NM_001322811.1 | NP_001309740.1 | 5 | 1424 | 67 | ||
| GenBank mRNA | AK130527.1 | - | 4 | 1891 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30328000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30327992 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30327991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30327990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30328005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30328010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30328013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30328001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09823772 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 414919 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 70578 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151985 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 137589 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 37894 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3221971 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0999 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0998 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 59368 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03082553 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3221971 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3221971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328013 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823773 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30327992 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30327990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30327992 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30327990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823773 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328013 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30328004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09823796 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09823765 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09823766 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09823761 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09823768 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16825997 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09823768 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03082548 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03082546 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03082548 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33788435 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20343782 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P451406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823781 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823841 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823818 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823776 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823827 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823839 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823826 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823825 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823801 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823783 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823803 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823820 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823779 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823824 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823819 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823775 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823833 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823836 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823785 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823794 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823772 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823817 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823788 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823837 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823796 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16826011 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823797 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823811 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823831 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823792 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823822 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823805 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823777 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823813 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09823809 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03082548 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001013657_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7985960 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7985962 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7985961 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7991394 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7991392 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7991393 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7985959 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7991386 | - | - |
| Affymetrix Human Genome U219 Array | 11735524_at | - | - |
| Affymetrix Human Genome U219 Array | 11735524_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053231 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053231 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_16995-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4860474 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_16995 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1810855 | AK130527.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1810855 | NM_001013657.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1810855 | AK130527.1 | human_transcriptome |

GDPGP1
Genomic Map