Assay Details
| Gene Symbol: MAN2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026818.1 | XP_016882307.1 | 5 - 6 | 762 | 89 | |
| GenBank mRNA | U68382.1 | - | 7 - 8 | 1005 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017026818.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

MAN2B1
Genomic Map